missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRAT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116166
This item is not returnable.
View return policy
Description
FRAT2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this intronless gene belongs to the GSK-3-binding protein family. Studies show that this protein plays a role as a positive regulator of the WNT signaling pathway. It may be upregulated in tumor progression.
Specifications
| FRAT2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O75474 | |
| FRAT2 | |
| KLH conjugated synthetic peptide between 203-230 amino acids from the C-terminal region of human FRAT2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| FRAT2 | |
| FRAT2; FRAT-2; frequently rearranged in advanced T cell lymphomas 2; frequently rearranged in advanced T-cell lymphomas 2; GSK-3; GSK-3 binding protein FRAT2; GSK-3-binding protein FRAT2; MGC10562; RP11-452K12.2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 23401 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction