missing translation for 'onlineSavingsMsg'
Learn More

FRAT2 Antibody [DyLight 405], Novus Biologicals™

Product Code. 30734195 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734195 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734195 Supplier Novus Biologicals Supplier No. AF7980E

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Sheep Polyclonal Antibody

FRAT2 Polyclonal antibody specifically detects FRAT2 in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen FRAT2
Applications Immunocytochemistry
Classification Polyclonal
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias frequently rearranged in advanced T-cell lymphomas 2FRAT-2, GSK-3 binding protein FRAT2, GSK-3-binding protein FRAT2, MGC10562
Host Species Sheep
Immunogen E. coli-derived recombinant human FRAT2, Ser31-Leu233 (Ala83Thr), Accession # O75474
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 23401
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.