missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529145
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene belongs to the GSK-3-binding protein family. The protein inhibits GSK-3-mediated phosphorylation of beta-catenin and positively regulates the Wnt signaling pathway. It may function in tumor progression and in lymphomagenesis. [provided by RefSeq, Oct 2008].
Specifications
| FRAT1 | |
| Polyclonal | |
| Unconjugated | |
| FRAT1 | |
| AW060382; Frat1; FRAT-1; frequently rearranged in advanced T cell lymphomas; frequently rearranged in advanced T-cell lymphomas 1; Proto-oncogene FRAT1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10023, 14296 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P70339, Q92837 | |
| FRAT1 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FRAT1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction