missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRAT1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33265-20ul
This item is not returnable.
View return policy
Description
FRAT1 Monoclonal antibody specifically detects FRAT1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunocytochemistry/Immunofluorescence
Specifications
| FRAT1 | |
| Monoclonal | |
| ELISA Recommended starting concentration is 1 μg/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| FLJ97193, FRAT-1, frequently rearranged in advanced T-cell lymphomas, Frequently rearranged in advanced T-cell lymphomas 1, proto-oncogene FRAT1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-170 of human FRAT1 (NP_005470.2).,, Sequence:, PLRAPGPLAAAVPADKARSPAVPLLLPPALAETVGPAPPGVLRCALGDRGRVRGRAAPYCVAELATGPSALSPLPPQADLDGPPGAGKQGIPQPLSGPCRR | |
| 20 μL | |
| Wnt Signaling Pathway | |
| 10023 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction