missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FRAS1 Related Extracellular Matrix 3 Polyclonal specifically detects FRAS1 Related Extracellular Matrix 3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | FRAS1 Related Extracellular Matrix 3 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | P0C091 |
| Gene Alias | FRAS1-Related Extracellular Matrix Protein 3, FREM3 |
| Gene Symbols | FRAS1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?