missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRAS1 (Fraser syndrome 1) Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | FRAS1 (Fraser syndrome 1) |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18156988
|
Novus Biologicals
NBP2-38656 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637485
|
Novus Biologicals
NBP2-38656-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FRAS1 (Fraser syndrome 1) Polyclonal specifically detects FRAS1 (Fraser syndrome 1) in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| FRAS1 (Fraser syndrome 1) | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| DKFZp686I05113, extracellular matrix protein FRAS1, FLJ14927, FLJ22031, Fraser syndrome 1, KIAA1500DKFZp686P08111 | |
| FRAS1 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q86XX4 | |
| 80144 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: IWIQLNYLPSYGTLLRISGSEVEELSEVSNFTMEDINNKKIRYSAVFETDGHLVTDSFYFSVSDMDHNHLDNQIFTIMITPAENPPPVIAFADLITVDEGGRAPLSFHHFFATDDDDNLQRDAIIKLSALPKYGCIENTGTGDRFG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title