Learn More
Fisherbrand™ Frame for IEF unit FB58120 glass electrode
Brand: Fisherbrand™
Code : 46
Additional Details : Weight : 1.00000kg
Description
The unit operates in one of three modes: wick based electrophoresis; pre-cast horizontal gels; isoelectric focusing.Design features incorporate:
- Robust acrylic construction
- Safety lid with integral power leads
- Shrouded power connectors
- Long life 99.99% pure replaceable platinum electrodes
- Gold plate terminal with stainless steel washers to resist corrosion
- Printed grid - permits migration rates to be measured
- Integral cooling coil - allows for either a recirculating chiller or for a water connection to chill the ceramic plate to the desired temperature
- Large cooled ceramic top plate - provides uniform cooling of the gel platform
- Two sets of electrodes: Tank mounted - these are used for running “wick” gels using the lower buffer chambers Detachable - clip onto one of two frames for either IEF or horizontal pre-cast gel running
- Top frame options (both with variable electrode spacing) Hollow frame - for IEF where the strips are placed directly onto the ceramic plate Glass bottomed frame - for pre-cast horizontal gels
- Buffer drainage ports. The inbuilt shut-off valve is opened when the drainage tube is connected to empty the buffer chamber