missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FRA7G(D7S486)/CEN7q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
£1235.00 - £2030.00
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | fragile site, aphidicolin type, common, fra(7)(q31.2) |
Gene Alias | D7S486 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16003345
|
Abnova™
FG0211.200uL |
200 μL |
£2030.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16093335
|
Abnova™
FG0211.100uL |
100 μL |
£1235.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
D7S486 | |
2414 | |
Liquid |
Tube | |
fragile site, aphidicolin type, common, fra(7)(q31.2) | |
FRA7G | |
DNA/RNA |