missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fra2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110687
This item is not returnable.
View return policy
Description
Immunogen sequence: EKLEFMLVAH GPVCKISPEE RRSPPAPGLQ PMRSGGGSVG AVVVKQEPLE EDSPSSSSAG LDKAQRSVIK PISIAGGFYG EEPLHTPIVV TSTPAVTPGT SNLVFTYPSV LEQESPASPS ESCSKAHRRS SSSGDQSSDS L Highest antigen sequence indentity to the following orthologs: Mouse - 88%, Rat - 58%.
The Fos gene family consists of 4 members: FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation.
Specifications
| Fra2 | |
| Polyclonal | |
| Unconjugated | |
| FOSL2 | |
| FOS like 2, AP-1 trancription factor subunit; FOS like 2, AP-1 transcription factor subunit; FOS like antigen 2; FOSL2; FOS-like antigen 2; fos-related antigen 2; Fra2; FRA-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14284, 2355, 25446 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.30 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P15408, P47930, P51145 | |
| FOSL2 | |
| Recombinant Protein corresonding to Human Fra2. Recombinant protein control fragment (Product #RP-102469). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction