missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA2 Antibody (PCRP-FOSL2-1B1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08467AF750
This item is not returnable.
View return policy
Description
FRA2 Monoclonal antibody specifically detects FRA2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| FRA2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| FOS-like antigen 2, fos-related antigen 2, FRA-2, FRA2FLJ23306 | |
| Recombinant full-length human FRA2 protein (Uniprot: P15408) | |
| 100 μg | |
| Cancer, Transcription Factors and Regulators, Tumor Suppressors | |
| 2355 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunofluorescence, Protein Array | |
| PCRP-FOSL2-1B1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction