missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fra1 Recombinant Rabbit Monoclonal Antibody (8M3Q2)

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557898

 View more versions of this product

Product Code. 30283074

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MFRDFGEPGP SSGNGGGYGG PAQPPAAAQA AQQKFHLVPS INTMSGSQEL QWMVQPHFLG PSSYPRPLTY PQYSPPQPRP GVIRALGPPP GVRRRPCEQI.

The v-Fos oncogene was initially detected in two independent murine osteosarcoma virus isolates and an avian nephroblastoma virus. Members of the c-Fos gene family, including c-Fos, Fos B, Fra-1 and Fra-2, encode nuclear phosphoproteins that are rapidly and transiently induced by a variety of agents and function as transcriptional regulators for several genes. In contrast to c-Jun proteins, which form homo- and heterodimers that bind to specific DNA response elements, c-Fos proteins are only active as heterodimers with members of the Jun gene family. In addition, selected ATF/CREB family members can form leucine zipper dimers with Fos and Jun. Different dimers exhibit differential specificity and affinity for AP-1 and CRE sites.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fra1
Recombinant Monoclonal
1.24 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P15407
FOSL1
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FRA1 (NP_005429.1).
100 μL
Primary
Human
Antibody
IgG
ELISA, Western Blot
8M3Q2
Unconjugated
FOSL1
AW538199; FOS like 1, AP-1 trancription factor subunit; FOS like 1, AP-1 transcription factor subunit; FOS like antigen 1; Fosl1; FOS-like antigen 1; FOS-like antigen-1; fos-related antigen 1; FRA; Fra1; fra-1; HGNC:13718; LOW QUALITY PROTEIN: fos-related antigen 1
Rabbit
Protein A
RUO
8061
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.