missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fra1 Recombinant Rabbit Monoclonal Antibody (8M3Q2)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557897
This item is not returnable.
View return policy
Description
Immunogen sequence: MFRDFGEPGP SSGNGGGYGG PAQPPAAAQA AQQKFHLVPS INTMSGSQEL QWMVQPHFLG PSSYPRPLTY PQYSPPQPRP GVIRALGPPP GVRRRPCEQI.
The v-Fos oncogene was initially detected in two independent murine osteosarcoma virus isolates and an avian nephroblastoma virus. Members of the c-Fos gene family, including c-Fos, Fos B, Fra-1 and Fra-2, encode nuclear phosphoproteins that are rapidly and transiently induced by a variety of agents and function as transcriptional regulators for several genes. In contrast to c-Jun proteins, which form homo- and heterodimers that bind to specific DNA response elements, c-Fos proteins are only active as heterodimers with members of the Jun gene family. In addition, selected ATF/CREB family members can form leucine zipper dimers with Fos and Jun. Different dimers exhibit differential specificity and affinity for AP-1 and CRE sites.
Specifications
| Fra1 | |
| Recombinant Monoclonal | |
| 1.24 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P15407 | |
| FOSL1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human FRA1 (NP_005429.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8M3Q2 | |
| Unconjugated | |
| FOSL1 | |
| AW538199; FOS like 1, AP-1 trancription factor subunit; FOS like 1, AP-1 transcription factor subunit; FOS like antigen 1; Fosl1; FOS-like antigen 1; FOS-like antigen-1; fos-related antigen 1; FRA; Fra1; fra-1; HGNC:13718; LOW QUALITY PROTEIN: fos-related antigen 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8061 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering