missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRA-1/FOSL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£343.00 - £497.00
Specifications
| Antigen | FRA-1/FOSL1 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18363086
|
Novus Biologicals
NBP3-17851-25UL |
25 μg |
£343.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18302157
|
Novus Biologicals
NBP3-17851-100UL |
100 μg |
£497.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FRA-1/FOSL1 Polyclonal antibody specifically detects FRA-1/FOSL1 in Human samples. It is validated for ImmunofluorescenceSpecifications
| FRA-1/FOSL1 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Wnt Signaling Pathway | |
| PBS, pH 7.2, 40% glycerol | |
| 8061 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FOS-like antigen 1, FOS-like antigen-1, fos-related antigen 1, FRA, FRA1, FRA-1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LVPSINTMSGSQELQWMVQPHFLGPSSYPRPLTYPQYSPPQPRPGVIRALGPPPGVRRRPCEQISPE | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title