missing translation for 'onlineSavingsMsg'
Learn More

FR 900098 Monosodium Salt, TRC

Product Code. 30395824
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395824 25 mg -
30402988 5 mg -
30394416 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395824 Supplier Toronto Research Chemicals Supplier No. TRCF76350025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73226-73-0
Molecular Formula C5 H11 N O5 P . Na
Molecular Weight (g/mol) 219.11
Synonym Phosphonic acid, P-[3-(acetylhydroxyamino)propyl]-, sodium salt (1:1), Phosphonic acid, [3-(acetylhydroxyamino)propyl]-, monosodium salt (9CI), FR 900098 monosodium salt, P-[3-(Acetylhydroxyamino)propyl]phosphonic acid sodium salt, (3-(N-Hydroxyacetamido)propyl)phosphonic acid sodium salt, [3-(Acetylhydroxyamino)propyl]phosphonic acid sodium salt
IUPAC Name sodium;3-[acetyl(hydroxy)amino]propyl-hydroxyphosphinate
SMILES [Na+].CC(=O)N(O)CCCP(=O)(O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C5H12NO5P.Na/c1-5(7)6(8)3-2-4-12(9,10)11;/h8H,2-4H2,1H3,(H2,9,10,11);/q;+1/p-1
Formula Weight 219.0273
Percent Purity >95%
Chemical Name or Material FR 900098 Monosodium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.