missing translation for 'onlineSavingsMsg'
Learn More

FR-188582, MedChemExpress

Product Code. 16472398
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472398 5 mg 5mg
16470199 10 mg 10mg
16450716 20 mg 20mg
16413189 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472398 Supplier Medchem Express Supplier No. HYU00146
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FR-188582 is a highly selective inhibitor of cyclooxygenase (COX)-2, with an IC50 value of 17 nM.

FR-188582 is a highly selective inhibitor of cyclooxygenase (COX)-2, with an IC50 value of 17 nM.

Specifications

Chemical Name or Material FR-188582
CAS 189699-82-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C16H13ClN2O2S
Quantity 5 mg
SMILES O=S(C1=CC=C(C2=CC(Cl)=NN2C3=CC=CC=C3)C=C1)(C)=O
Molecular Weight (g/mol) 332.8
Formula Weight 332.8
Percent Purity 99.21%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.