missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPRL2/FPR3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26058
This item is not returnable.
View return policy
Description
FPRL2/FPR3 Polyclonal specifically detects FPRL2/FPR3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| FPRL2/FPR3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FPR3 | |
| Peptide with sequence C-DSAQTSNTDTTS corresponding to C-Terminus according to NP_002021.3. | |
| 0.1 mg | |
| GPCR, Immunology | |
| 2359 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1:100 - 1:2000, Peptide ELISA Detection limit 1:1000 | |
| FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction