missing translation for 'onlineSavingsMsg'
Learn More

FPRL1/FPR2 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18351467 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18351467 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18351467 Supplier Novus Biologicals Supplier No. NLS1878R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FPRL1/FPR2 Polyclonal specifically detects FPRL1/FPR2 in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FPRL1/FPR2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:300, Immunocytochemistry/Immunofluorescence 1:20-1:75, Immunohistochemistry-Paraffin 1:300, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P25090
Gene Alias ALXR, FMLP-R-II, FMLPX, formyl peptide receptor 2, Formyl peptide receptor-like 1RFP, FPR2A, FPRH1FMLP-related receptor I, FPRL1LXA4 receptor, HM63FPRH2, Lipoxin A4 receptor, lipoxin A4 receptor (formyl peptide receptor related), LXA4RFMLP-R-I, N-formyl peptide receptor 2
Gene Symbols FPR2
Host Species Rabbit
Immunogen This FPRL1/FPR2 antibody is made to a synthetic peptide made to the 2nd extracellular loop of the human FPRL1 protein (between residues 300-350) [UniProt P25090]
Molecular Weight of Antigen 38 kDa
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2358
Test Specificity Human and mouse. Rat reactivity reported in scientific literature (PMID: 24086560).
Target Species Human, Mouse, Rat, Bacteria
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.