missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: DTYCTFNFAS WGGTPEERLK VAITMLTARG Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
Formyl peptide receptor 2 (FPR2) is a low affinity receptor for N-formyl-methionyl peptides. Binding of FMLP to FPR2 causes activation of neutrophils. The response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. The activation of LXA4R could result in an anti-inflammatory outcome counteracting the actions of proinflammatory signals such as LTB4.
Specifications
Specifications
| Antigen | FPR2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | FPR2 |
| Gene Accession No. | P25090 |
| Gene Alias | ALXR; E330010I07Rik; fMLPR1; FMLP-related receptor I; FMLP-R-I; FMLP-R-II; FMLPX; formyl peptide receptor 2; formyl peptide receptor, related sequence 2; formyl peptide receptor, related sequence 2-like; formyl peptide receptor-like 1; Formylpeptide receptor-related sequence 2; FPR2; FPR2A; FPRH1; FPRH2; Fprl1; Fprl-1; FPR-like 1; Fpr-rs1; Fpr-rs2; HM63; Lipoxin A4 receptor; lipoxin A4 receptor (formyl peptide receptor related); lipoxin A4 receptor-like protein; Lipoxin receptor; LXA4 receptor; LXA4R; N-formyl peptide receptor 2; N-formylpeptide receptor-like 2; RFP |
| Gene Symbols | FPR2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
