missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FPR Agonist 43, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
FPR Agonist 43 (compound 43) is a dual formyl peptide receptor 1 (FPR1) and formyl peptide receptor 2 (FPR2)/ALX agonist.
Specifications
Specifications
| Chemical Name or Material | FPR Agonist 43 |
| Physical Form | Powder |
| CAS | 903895-98-7 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H21ClN4O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 125 mg/mL (324.79 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C(NC1=C(C(C)C)N(C)N(C2=CC=CC=C2)C1=O)NC3=CC=C(Cl)C=C3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction