missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FPGT Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119127
This item is not returnable.
View return policy
Description
FPGT Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
L-fucose is a key sugar in glycoproteins and other complex carbohydrates since it may be involved in many of the functional roles of these macromolecules, such as in cell-cell recognition. The fucosyl donor for these fucosylated oligosaccharides is GDP-beta-L-fucose. There are two alternate pathways for the biosynthesis of GDP-fucose; the major pathway converts GDP-alpha-D-mannose to GDP-beta-L-fucose. The protein encoded by this gene participates in an alternate pathway that is present in certain mammalian tissues, such as liver and kidney, and appears to function as a salvage pathway to reutilize L-fucose arising from the turnover of glycoproteins and glycolipids. This pathway involves the phosphorylation of L-fucose to form beta-L-fucose-1-phosphate, and then condensation of the beta-L-fucose-1-phosphate with GTP by fucose-1-phosphate guanylyltransferase to form GDP-beta-L-fucose.
Specifications
| FPGT | |
| Polyclonal | |
| Unconjugated | |
| FPGT | |
| 1700016E03Rik; Fpgt; fucose-1-phosphate guanyltransferase; fucose-1-phosphate guanylyltransferase; GDP-beta-L-fucose pyrophosphorylase; GDP-L-fucose diphosphorylase; GDP-L-fucose pyrophosphorylase; GFPP | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 75540, 8790 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| G5E8F4, O14772 | |
| FPGT | |
| Recombinant protein Fucose-1-Phosphate Guanylyltransferase. The antigen corresponds to amino acid range 314-513 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction