missing translation for 'onlineSavingsMsg'
Learn More

FP-Biotin (Mixture of Diastereomers), TRC

Product Code. 30395502
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395502 10 mg -
30392398 2.5 mg 2.5mg
30401376 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395502 Supplier Toronto Research Chemicals Supplier No. TRCF75540010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259270-28-5
Molecular Formula C27H50FN4O5PS
Molecular Weight (g/mol) 592.75
Synonym [10-[[5-[[5-[(3aS,4S,6aR)-Hexahydro-2-oxo-1H-thieno[3,4-d]imidazol-4-yl]-1-oxopentyl]amino]pentyl]amino]-10-oxodecyl]phosphonofluoridic Acid Ethyl Ester, 10-Fluoroethoxyphosphinyl-N-biotinamidopentyldecanamide
IUPAC Name ethyl (10-oxo-10-((5-(5-((3aS,4S,6aR)-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanamido)pentyl)amino)decyl)phosphonofluoridate
SMILES O=C(N1)N[C@@]2([H])[C@]1([H])CS[C@H]2CCCCC(NCCCCCNC(CCCCCCCCCP(OCC)(F)=O)=O)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H50FN4O5PS/c1-2-37-38(28,36)20-14-7-5-3-4-6-9-16-24(33)29-18-12-8-13-19-30-25(34)17-11-10-15-23-26-22(21-39-23)31-27(35)32-26/h22-23,26H,2-21H2,1H3,(H,29,33)(H,30,34)(H2,31,32,35)/t22-,23-,26-,38?/m0/s1
Formula Weight 592.32
Percent Purity >95%
Chemical Name or Material FP-Biotin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.