missing translation for 'onlineSavingsMsg'
Learn More

FOY 251, TRC

Product Code. 17963795
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963795 5 mg 5mg
16645613 10 mg 10mg
17933445 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17963795 Supplier Toronto Research Chemicals Supplier No. TRCF7545005MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71079-09-9
Molecular Formula C16 H15 N3 O4 . C H4 O3 S
Molecular Weight (g/mol) 409.41
IUPAC Name 2-[4-(4-carbamimidamidobenzoyl)oxyphenyl]acetic acid;methanesulfonic acid
SMILES CS(=O)(=O)O.NC(=N)Nc1ccc(cc1)C(=O)Oc2ccc(CC(=O)O)cc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C16H15N3O4.CH4O3S/c17-16(18)19-12-5-3-11(4-6-12)15(22)23-13-7-1-10(2-8-13)9-14(20)21;1-5(2,3)4/h1-8H,9H2,(H,20,21)(H4,17,18,19);1H3,(H,2,3,4)
Formula Weight 409.0944
Chemical Name or Material FOY 251
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.