missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16234-1-AP-20UL
This item is not returnable.
View return policy
Description
The forkhead family of transcription factors belongs to the winged helix class of DNA-binding proteins. The protein encoded by this intronless gene contains a forkhead domain and is found predomitly in aorta and kidney. The function of the encoded protein is unknown.Specifications
| FOXS1 | |
| Polyclonal | |
| Unconjugated | |
| FOXS1 | |
| FKHL18, Forkhead box protein S1, forkhead box S1, Forkhead like 18 protein, FOXS1, FREAC 10, FREAC10 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14239, 2307, 311547 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43638, Q61574 | |
| Foxs1 | |
| FOXS1 Fusion Protein Ag8957 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction