missing translation for 'onlineSavingsMsg'
Learn More

FOXRED2 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Product Code. 18418085 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418085 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418085 Supplier Novus Biologicals Supplier No. NBP281943AF594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FOXRED2 Polyclonal antibody specifically detects FOXRED2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXRED2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias Endoplasmic reticulum flavoprotein associated with degradation, ERFAD, FAD-dependent oxidoreductase domain containing 2, FAD-dependent oxidoreductase domain-containing protein 2, FLJ23322, FLJ33734
Host Species Rabbit
Immunogen FOXRED2 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human FOXRED2. The immunogen is located within amino acids 70 - 120 of FOXRED2. Amino Acid Squence: SINKRYTGKANAEFNLR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80020
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.