Learn More
FOXRED1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565242
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: LSVGGIRQQFS LPENIQLSLF SASFLRNINE YLAVVDAPPL DLRFNPSGYL LLASEKDAAA MESNVKVQRQ EGAKVSLMSP DQLRNK Highest antigen sequence identity to the following orthologs - mouse 79%, rat 80%.
This gene encodes a protein that contains a FAD-dependent oxidoreductase domain. The encoded protein is localized to the mitochondria and may function as a chaperone protein required for the function of mitochondrial complex I. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants have been observed for this gene.Specifications
FOXRED1 | |
Polyclonal | |
Unconjugated | |
FOXRED1 | |
BC024806; FAD dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain containing 1; FAD-dependent oxidoreductase domain-containing protein 1; Foxred1; FP634; H17; RGD1311785; TEG-23; testis expressed gene 23; Tex23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55572 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CU9 | |
FOXRED1 | |
Recombinant Human FOXRED1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only