missing translation for 'onlineSavingsMsg'
Learn More

FOXR1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

£359.00

Specifications

Antigen FOXR1
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18363244
View Documents
Novus Biologicals
NBP3-09247-100UL
100 μg
£359.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FOXR1 Polyclonal specifically detects FOXR1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

FOXR1
Western Blot
Unconjugated
Rabbit
Human
DLNB13, Forkhead box protein N5, forkhead box R1, forkhead box R1 variant 1, FOXN5forkhead box protein R1, MGC149486
The immunogen is a synthetic peptide directed towards the N terminal region of human FOXR1 (NP_859072). Peptide sequence KKPNPDKDGPDYEPNLWMWVNPNIVYPPGKLEVSGRRKREDLTSTLPSSQ
Affinity purified
Western Blot 1.0 ug/ml
Polyclonal
Purified
RUO
PBS buffer, 2% sucrose
283150
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.