missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXR1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144774
This item is not returnable.
View return policy
Description
Immunogen sequence: LPSPHKRAPL QSRRLRQASS QAGRLWSRPP LNYFHLIALA LRNSSPCGLN VQQIYSFTRK HFPFFRTAPE GWKNTVRHNL CFRDSFEKVP VSMQGGASTR PRSCLWKLTE EGHRRFAEEA RALASTRLES IQQCMSQPD Antigen sequence identity: Mouse 54%; Rat 84%.
The forkhead-box (FOX) genes comprise a superfamily of at least 43 members that express proteins which are involved in transcriptional regulation and may be associated with the pathogenesis of various cancers. FOX family members are monomeric, helix-turn-helix proteins with a core DNA-binding domain of approximately 110 amino acids. FOX transcription factors play roles in determining cell fate during early development. FOXR1 (forkhead box protein R1), also known as FOXN5 (forkhead box protein N5) or DLNB13, is a 292 amino acid nuclear protein that belongs to the FOX family and contains one forkhead DNA-binding domain. Through chromosomal aberrations such as retroviral integration, gene amplification or translocation, FOXR1 may be involved the development of certain invasive carcinomas.
Specifications
| FOXR1 | |
| Polyclonal | |
| Unconjugated | |
| Foxr1 | |
| DLNB13; Forkhead box protein N5; forkhead box protein R1; forkhead box R1; forkhead box R1 variant 1; forkhead box R1 variant 2; Foxn5; FOXR1; Gm1115 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 283150 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PIV2 | |
| Foxr1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 146-284. Recombinant protein control fragment (Product #RP-109716). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction