missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXQ1 Monoclonal Antibody (OTI4D9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526943
This item is not returnable.
View return policy
Description
FOXQ1 Monoclonal Antibody for Western Blot, IHC (P)
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
Specifications
| FOXQ1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9C009 | |
| FOXQ1 | |
| Human recombinant protein fragment corresponding to amino acids 11-295 of FOXQ1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4D9 | |
| Unconjugated | |
| FOXQ1 | |
| forkhead box protein Q1; forkhead box Q1; FOXQ1; Hepatocyte nuclear factor 3 forkhead homolog 1; Hfh1; HFH-1; Hfh1l; HFH-1l; HNF-3/forkhead homolog 1 like; HNF-3/forkhead homolog-1; HNF-3/forkhead-like protein 1; sa; satin; winged helix/forkhead transcription factor | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 94234 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction