missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXQ1 Monoclonal Antibody (2F2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA522893
This item is not returnable.
View return policy
Description
Peptide Sequence: RSKPYTRRPK PPYSYIALIA MAIRDSAGGR LTLAEINEYL MGKFPFFRGS YTGWRNSVRH NLSLNDCFVK VLRDPSRPWG KDNYWMLNPN SEYTFADGVF RRRRKRLSHR.
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
Specifications
| FOXQ1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q9C009 | |
| FOXQ1 | |
| FOXQ1 (NP_150285, 110 a.a. ~ 219 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry | |
| 2F2 | |
| Unconjugated | |
| FOXQ1 | |
| forkhead box protein Q1; forkhead box Q1; FOXQ1; Hepatocyte nuclear factor 3 forkhead homolog 1; Hfh1; HFH-1; Hfh1l; HFH-1l; HNF-3/forkhead homolog 1 like; HNF-3/forkhead homolog-1; HNF-3/forkhead-like protein 1; sa; satin; winged helix/forkhead transcription factor | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 94234 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction