missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXQ1 Antibody (OTI4D9), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72338CL1
This item is not returnable.
View return policy
Description
FOXQ1 Monoclonal antibody specifically detects FOXQ1 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot
Specifications
| FOXQ1 | |
| Monoclonal | |
| CoraFluor 1 | |
| forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor | |
| Human recombinant protein fragment corresponding to amino acids 11-295 of human FOXQ1(NP_150285) produced in E.coli. | |
| 0.1 mL | |
| Cancer | |
| 94234 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| OTI4D9 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction