missing translation for 'onlineSavingsMsg'
Learn More

FOXQ1 Antibody (2F2), Novus Biologicals™

Product Code. 18403899 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403899 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403899 Supplier Novus Biologicals Supplier No. H00094234M05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FOXQ1 Monoclonal antibody specifically detects FOXQ1 in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXQ1
Applications ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 2F2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_150285
Gene Alias forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor
Host Species Mouse
Immunogen FOXQ1 (NP_150285, 110 a.a. ∽ 219 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RSKPYTRRPKPPYSYIALIAMAIRDSAGGRLTLAEINEYLMGKFPFFRGSYTGWRNSVRHNLSLNDCFVKVLRDPSRPWGKDNYWMLNPNSEYTFADGVFRRRRKRLSHR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 94234
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.