missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXP4, Mouse anti-Human, Clone: 3B12, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00116113-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: LNSPGMLNPGSASSLLPLSHDDVGAPVEPLPSNGSSSPPRLSPPQYSHQVQVKEEPAEAEEDRQPGPPLGAPNPSASGPPEDRDLEEELPGEELSpecifications
| FOXP4 | |
| Monoclonal | |
| Unconjugated | |
| forkhead box P4 | |
| FLJ40908/FLJ44184/hFKHLA | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3B12 | |
| In 1x PBS, pH 7.4 | |
| NM_001012426 | |
| FOXP4 | |
| FOXP4 (NP_001012426.1, 586 a.a. ∼ 679 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 116113 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction