missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXP4 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-S40)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548342
This item is not returnable.
View return policy
Description
Specificity: This antibody recognizes FOXP4 (Forkhead box protein P4). It binds to a folded domain, amino acids 302-541. FOXP4 is a transcriptional repressor that represses lung-specific expression.
This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Many members of the forkhead box gene family, including members of subfamily P, have roles in mammalian oncogenesis. This gene may play a role in the development of tumors of the kidney and larynx. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
Specifications
| FOXP4 Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q8IVH2 | |
| Foxp4 | |
| This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP4 protein under non-denaturing conditions. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| RAB-S40 | |
| Unconjugated | |
| Foxp4 | |
| 1200010K03Rik; 2310007G05Rik; FKHLA; FLJ40908; FLJ44184; fork head-related protein like A; fork head-related protein-like A; forkhead box P4; forkhead box protein P4; Foxp4; hFKHLA; mFKHLA; OTTHUMP00000043536; winged-helix repressor FOXP4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 116113 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction