missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Foxp3 / Transcription Factor Staining Buffer Set

Product Code. 11500597
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11500597 1 kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains Sodium azide and formaldehyde
Product Code. 11500597 Supplier Invitrogen™ Supplier No. 00552300
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Contains Sodium azide and formaldehyde

Formulated and optimized for staining with antibodies against transcription factors and nuclear proteins (e.g. Foxp3 and Ki-67), cytokines and chemokines

The Foxp3 Transcription Factor Staining Buffer Set has been formulated and optimized for staining with antibodies to transcription factors and nuclear proteins, such as Foxp3 and Ki-67, as well as cytokines and chemokines.

For your convenience, we have provided these protocols for preparing your samples for intracellular antigen detection on a flow cytometer.

Reported Application
Intracellular Staining Followed by Flow Cytometric Analysis

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 kit
Reagent Type Fixation Diluent, Fixation Reagent, Permeabilization Diluent, Permeabilization Reagent
Product Line eBioscience
Product Type Staining Buffer
compliance-icons compliance-icons
Product Identifier
  • Foxp3 / Transcription Factor Staining Buffer Set
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Formaldehyde, Methanol

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.