missing translation for 'onlineSavingsMsg'
Learn More

Anti-Mouse/Rat Foxp3 Staining Set PE, eBioscience™

Antibody Staining Kit

Brand:  Invitrogen 72-5775-40

Product Code. 15580707

  • £346.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • Fixation/Permeabilization Concentrate: 30mL (4X). Store at 2-8°C. Avoid agitation. Dilute 1 part Fixation/Permeabilization Concentrate with 3 parts Fixation/Permeabilization Diluent to make the Fixation/Permeabilization working solution. Caution: This solution contains Paraformaldehyde, which is toxic and a suspected carcinogen. Contact with eyes, skin and mucous membranes should be avoided. Wear proper protective clothing and gloves.
  • Fixation/Permeabilization Diluent: 100mL. Store at 2-8°C. The diluent is intended to be used in combination with the Fixation/Permeabilization Concentrate.
  • Permeabilization Buffer (10X): 100mL. Store at 2-8°C. Dilute to 1X with deionized/distilled water and store at 4°C. Caution: Harmful if swallowed or irritant by contact. Wear proper protective clothing and gloves.
  • anti-mouse/rat Foxp3 (FJK-16s): 25μg. Store at 2-8°C in the dark.
This item is not returnable. View return policy

Description

Description

The FJK-16s antibody reacts with mouse, rat, dog, porcine, and bovine Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in ‘scurfy’ (sf) mice. Constitutive high expression of foxP3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

Immunoblotting with FJK-16s antibody has mapped the epitope to amino acids 75-125 of the mouse Foxp3 protein. In the human, this region has been shown to be alternatively spliced at the mRNA level. Both the alternatively-spliced and non-spliced isoforms are present in the CD4+CD25+ subset of lymphocytes. Preliminary RT-PCR experiments have not revealed this alternatively-spliced isoform in mouse splenocytes, suggesting different gene regulation in the mouse and human.

Intracellular staining of mouse splenocytes with FJK-16s using the PE anti-mouse/rat Foxp3 Staining Set and protocol reveals approximately 2% of total cells in the C57Bl/6 strain and approximately 3-5% in the BALB/c mouse strain. Multicolor flow cytometric analysis demonstrates approximately 90% of the CD4+CD25+ cells and 4% of the CD4+CD25- cells staining with FJK-16s. B220+, CD11b+, CD11c+, and Ly6G/Gr-1+ cells do not show significant co-staining with FJK-16s.

Please see our FAQ regarding the usage of eBioscience Foxp3 reagents.

Not included:
Fc Block (cat. 14-0161)
Flow Cytometry Staining Buffer (cat. 00-4222)
Rat IgG2a isotype Control (cat. 12-4321, 11-4321, or 17-4321)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Foxp3
Monoclonal
0.2mg/mL
Rat
RUO
Mouse, Rat
IgG2a κ
Flow Cytometry
FJK-16s
PE
1 kit
Primary
Store at 2-8°C. Light sensitive material. Use within 6 months of opening or by date indicated on the bottle.
eBioscience™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.