missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXP3 Polyclonal Antibody, DyLight™ 550, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522776
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody has shown reactivity against Ms (CD4+CD25+ T cells), does not react with Hu Jurkat, Ms CD4+CD25- T cell lysates.
FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.Specifications
FOXP3 | |
Polyclonal | |
DyLight 550 | |
Foxp3 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Rabbit | |
Protein G | |
RUO | |
20371, 317382, 506053, 50943 | |
4° C, store in dark | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.67 mg/mL | |
50mM sodium borate with 0.05% sodium azide | |
Q99JB6, Q9BZS1 | |
Foxp3 | |
A peptide correpsonding to amino acids 43-100 of mouse FOXP3 | |
100 μL | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |