missing translation for 'onlineSavingsMsg'
Learn More

Anti-Human Foxp3 Staining Set APC, eBioscience™

Antibody Staining Kit

Brand:  Invitrogen 77-5774-40

Product Code. 15295842

  • £318.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!


Includes:
  • Fixation/Permeabilization Concentrate (4X) (cat. 00-5123): 30mL, store at 2-8°C. Avoid agitation.
  • Fixation/Permeabilization Diluent (cat. 00-5223): 100mL, store at 2-8°C.
  • Permeabilization Buffer (10X) (cat. 00-8333): 100mL, store at 2-8°C. Note: The 10X Permeabilization Buffer has a natural tendency to precipitate, however, its function is not affected by this. To clarify, the solution can be filtered after dilution to 1x working solution.
  • Normal Mouse Serum (cat. 24-5544): 100 μL, store at 2-8°C.
  • Anti-Human Foxp3 APC (clone 236A/E7, cat. 17-4777): 25 tests, store at 2-8°C. Light-sensitive material.
This item is not returnable. View return policy

Description

Description

This Anti-Human Foxp3 Staining Set contains the buffers and monoclonal antibody necessary to successfully stain and identify Foxp3+ cells. The 236A/E7 monoclonal antibody reacts with human Foxp3, also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3 is a 49-55kDa protein and a member of the forkhead/winged-helix family of transcription factors. It was identified as the gene responsible for the X-linked lymphoproliferative disease observed in scurfy (sf) mice and in the human disorder, X-linked autoimmunity-allergic dysregulation syndrome (XLAAD). Constitutive expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells.

The 236A/E7 antibody has been shown to crossreact with rhesus macaque, sooty mangabey, and cynomolgus Foxp3. 236A/E7 recognizes a different epitope of Foxp3 than PCH101 and 150D/E4.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Foxp3
Monoclonal
APC
Mouse
RUO
Cynomolgus, Human, Macaque, Monkey, Non-human Primate, Rhesus Monkey
IgG1
Flow Cytometry
236A/E7
Forkhead Box P3, Scurfin, JM2
1 kit
Primary
Store at 2-8°C. Light-sensitive material. Use within 6 months of opening or by date indicated on the bottle.
eBioscience
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.