Learn More
FOXP3 Monoclonal Antibody (NRRF-30), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 12-4771-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The NRRF-30 antibody reacts with mouse Foxp3 also known as FORKHEAD BOX P3, SCURFIN, and JM2; cross reactivity of this antibody to other proteins has not been determined. Foxp3, a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice. Constitutive high expression of foxP3 mRNA has been shown in CD4+/CD25+ regulatory T cells (Treg cells), and ectopic expression of foxp3 in CD4+/CD25- cells imparts a Treg phenotype in these cells. Immunoblotting with NRRF-30 antibody has mapped the epitope to amino acids 1-75 of the mouse Foxp3 protein. Intracellular staining of mouse splenocytes with fluorochrome-conjugated NRRF-30 using the eBioscience Foxp3 Staining Buffers (cat. 00-5523) and corresponding staining protocol reveals approximately 3% of total cells in the C57Bl/6 strain and approximately 5% in the BALB/c mouse strain. Multicolor flow cytometric analysis demonstrates approximately 90% of the CD4+/CD25+ cells and 4% of the CD4+/CD25- cells staining with NRRF-30. Co-staining with FJK-16s (anti-mouse/rat Foxp3 cat. 71-5775), which has been mapped to amino acids 71-125, and NRRF-30 shows 100% correlation, indicating that the same cells are stained with both anti-mouse Foxp3 antibodies. Please see our FAQ regarding the usage of eBioscience Foxp3 reagents.
FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.Specifications
FOXP3 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q99JB6 | |
Foxp3 | |
Affinity chromatography | |
RUO | |
20371 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
NRRF-30 | |
PE | |
Foxp3 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.