missing translation for 'onlineSavingsMsg'
Learn More

FoxP2 Antibody (1F8), Novus Biologicals™

Product Code. 18403299 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403299 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403299 Supplier Novus Biologicals Supplier No. H00093986M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

FoxP2 Monoclonal antibody specifically detects FoxP2 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxP2
Applications Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Sandwich ELISA
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055306
Gene Alias CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein
Host Species Mouse
Immunogen FOXP2 (NP_055306, 616 a.a. ∽ 715 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LAESSLPLLSNPGLINNASSGLLQAVHEDLNGSLDHIDSNGNSSPGCSPQPHIHSIHVKEEPVIAEDEDCPMSLVTTANHSPELEDDREIEEEPLSEDLE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93986
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.