missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxP1 Rabbit anti-Human, Clone: FOXP1/44R, Novus Biologicals™
Rabbit Monoclonal Antibody
£412.00
Specifications
| Antigen | FoxP1 |
|---|---|
| Clone | FOXP1/44R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
Description
FoxP1 Monoclonal specifically detects FoxP1 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| FoxP1 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ23741, fork head-related protein like B, forkhead box P1, forkhead box protein P1, glutamine-rich factor 1, hFKH1B, HSPC215, MGC12942, MGC88572, MGC99551, QRF112CC4 | |
| Synthetic peptide corresponding to residues within aa350-450 of FoxP1 | |
| Primary | |
| Store at 4°C. |
| FOXP1/44R | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 27086 | |
| IgG | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title