missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535117
Code : NEW
Additional Details : Weight : 0.01000kg
Description
FOXO6 is a transcriptional activator.Specifications
FOXO6 | |
Polyclonal | |
Unconjugated | |
FOXO6 | |
forkhead box O6; forkhead box protein O6; Foxo6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100132074, 329934 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
A8MYZ6, Q70KY4 | |
FOXO6 | |
KLH conjugated synthetic peptide between 154-181 amino acids from the central region of human FOXO6 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |