missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4, p Ser197 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£159.00 - £372.00
Specifications
| Antigen | FOXO4 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18646027
|
Novus Biologicals
NBP3-05393-100ul |
100 μg |
£372.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640647
|
Novus Biologicals
NBP3-05393-20ul |
20 μg |
£159.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FOXO4 Polyclonal antibody specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| FOXO4 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Phospho Specific | |
| PBS with 50% glycerol, pH7.3. | |
| 4303 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:100 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7 | |
| A synthetic phosphorylated peptide around S197 of human FOXO4 (NP_005929.2). AASMD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title