missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | FOXO3A |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817513
|
Proteintech
10849-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807513
|
Proteintech
10849-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The calculated molecular weight of FOXO3 is 71 kDa, but some experiments using a FOXO3a specific antibody showed the expression of a 90 kDa full-length FOXO3a protein (Full) and a 40 kDa processed FOXO3a product (Short).
FoxO (forkhead box O) proteins are a family of transcription factors characterized by the distinct forkhead domain. The FoxO proteins act in the insulin signaling pathway, but FoxO3 has also been shown to play a role in the regulation of apoptosis, cell cycle progression and cancer. Several splice variants of FoxO3 have been reported.Specifications
| FOXO3A | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| O43524, Q9WVH4 | |
| 2309, 294515, 56484 | |
| FOXO3A Fusion Protein Ag1289 | |
| Primary | |
| -20°C | |
| FOXO3 |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| AF6q21, AF6q21 protein, FKHRL1, FKHRL1P2, forkhead box O3, Forkhead box protein O3, FOXO2, FOXO3, FOXO3A | |
| Foxo3 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title