Learn More
Invitrogen™ FOXO3A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1805
Description
PA1-805 detects FoxO3. PA1-805 has been successfully used in Western blot procedures. In Western blot analysis of rat heart and lung, this antibody detects a ~ 71 kDa protein representing FoxO3. The PA1-805 immunogen is a synthetic peptide corresponding to residues L N(654) V G N F T G A K Q A S S Q S W V(670) of human FOXO3. This sequence is conserved in human, mouse, and rat.
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
Specifications
| FOXO3A | |
| Polyclonal | |
| Unconjugated | |
| FOXO3 | |
| 1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; C76856; Fkhr2; FKHRL1; FKHRL1P2; forkhead box O3; forkhead box O3a; forkhead box O3A transcription factor; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO2; FOXO3; FOXO-3; FOXO3A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2309, 294515, 56484 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| O43524, Q9WVH4 | |
| FOXO3 | |
| Synthetic peptide corresponding to residues L N(654) V G N F T G A K Q A S S Q S W V(670) of human FOXO3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.