missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FOXO3A Monoclonal Antibody (K.115.9)
GREENER_CHOICE

Product Code. 11544901
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11544901 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11544901 Supplier Invitrogen™ Supplier No. MA514932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay. It is not recommended to aliquot this antibody.

USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXO3A
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone K.115.9
Concentration 15 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene FOXO3
Gene Accession No. O43524, Q9WVH4
Gene Alias 1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; C76856; Fkhr2; FKHRL1; FKHRL1P2; forkhead box O3; forkhead box O3a; forkhead box O3A transcription factor; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO2; FOXO3; FOXO-3; FOXO3A
Gene Symbols FOXO3
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Glu50 of human FoxO3a.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2309, 294515, 56484
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.