missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO1A/3/4 Antibody (14E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-41195
This item is not returnable.
View return policy
Description
FOXO1A/3/4 Monoclonal specifically detects FOXO1A/3/4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| FOXO1A/3/4 | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 0.1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| FOXO1 | |
| Amino acids 242-655 of FKHR expressed in E. coli. | |
| 100 μL | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers | |
| 2308 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 140000 | |
| Unconjugated | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| This antibody will recognize human FKHR. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction