missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxO1/FKHR, p Ser256 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£213.00 - £341.00
Specifications
| Antigen | FoxO1/FKHR (p Ser256) |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217992
|
Novus Biologicals
NB100-81927-0.025ML |
0.025 mL |
£213.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18711614
|
Novus Biologicals
NB100-81927 |
£341.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
FoxO1/FKHR Polyclonal specifically detects FoxO1/FKHR in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| FoxO1/FKHR (p Ser256) | |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| FOXO1 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific | |
| Q12778 | |
| 2308 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title