missing translation for 'onlineSavingsMsg'
Learn More

FoxO1/FKHR Antibody, DyLight 488, Novus Biologicals™

Product Code. 18431976
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18431976 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18431976 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP224857G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FoxO1/FKHR Polyclonal antibody specifically detects FoxO1/FKHR in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxO1/FKHR
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma
Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human FOXO1A protein was used as the immunogen.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2308
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.