missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FoxO1/FKHR Antibody (83N7F8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-31376H
This item is not returnable.
View return policy
Description
FoxO1/FKHR Monoclonal specifically detects FoxO1/FKHR in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| FoxO1/FKHR | |
| Monoclonal | |
| HRP | |
| FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human. Based upon immunogen sequence simlarity, this antibody is predicted to react with Mouse (92%), Rat (92%), Bovine/Cow (91%), Squirrel (91%) and Porcine/Pig (90%). | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 83N7F8 | |
| Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| FOXO1 | |
| FOXO1A | |
| 0.1 mL | |
| Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific | |
| 2308 | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction