Learn More
Invitrogen™ FOXN3 Recombinant Rabbit Monoclonal Antibody (1H2L8)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 702764
Description
This antibody is predicted to react with Monkey, Horse, Cat Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.
Specifications
| FOXN3 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O00409 | |
| Foxn3 | |
| Peptides corresponding to human FOXN3 ICC/FC Sp (aa381-398, aa440-455). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ChIP Assay, Immunocytochemistry | |
| 1H2L8 | |
| Unconjugated | |
| Foxn3 | |
| 5430426H20Rik; AA589593; AW556347; C14orf116; checkpoint suppressor 1; CHES; Ches1; Ches1l; forkhead box N3; forkhead box protein N3; Foxn3; HTLFL1; PRO1635 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 1112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.